2-Aminofenol 99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

104260050 104261000 104265000
ACRO104260050EA 30.8 EUR
ACRO104260050 ACRO104261000 ACRO104265000
2-Aminofenol 99%
2-Aminofenol
Formule: C₆H₇NO
Kookpunt: 164 °C (15mbar)
Smeltpunt: 172…177 °C
Dichtheid: 1,328 g/cm³ (20 °C)
Vlampunt: 168 °C
MDL Nummer: MFCD00007690
CAS nummer: 95-55-6
EINECS: 202-431-1
UN: 2512
ADR: 6.1,III
Merck Index: 13,00460

Order Now

Specification Test Results

Appearance (Color) White to brown
Appearance (Form) Powder
Infrared spectrum Conforms
Melting point 172 to 177 °C
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR