α-Benzoinoxime 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

105530250 105531000 105535000
ACRO105530250EA 46.6 EUR
ACRO105530250 ACRO105531000 ACRO105535000
α-Benzoinoxime 98%
α-Benzoinoxime
Formule: C₁₄H₁₃NO₂
Smeltpunt: 150…155 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004501
CAS nummer: 441-38-3
EINECS: 207-127-2

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 150 to 155 °C
HPLC ≥97.5 %
GC ≥99.0 %
UV at 300 nm A: ≤0.01
Color scale ≤10 APHA
Residue after evaporation ≤0.002 %
Substance darkened by H2SO4 passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR