Phenylglyoxylic acid 97%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

105760050 105760250 105761000
ACRO105760050EA 29.1 EUR
ACRO105760050 ACRO105760250 ACRO105761000
Phenylglyoxylic acid 97%
Phenylglyoxylic acid
Formule: C₈H₆O₃
Smeltpunt: 60…69 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002575
CAS nummer: 611-73-4
EINECS: 210-278-7

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 60 to 69 °C
Titration with NaOH ≥96.0 %
GC ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR