α-Bromo-p-xylene 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

107570250 107570050 107571000
ACRO107570250EA 59.1 EUR
ACRO107570250 ACRO107570050 ACRO107571000
α-Bromo-p-xylene 98%
α-Bromo-p-xylene
Formule: C₈H₉Br
Kookpunt: 218…220 °C (760 mmHg)
Smeltpunt: 33…37 °C
Dichtheid: 1,324 g/cm³ (20 °C)
Vlampunt: 97 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00000181
CAS nummer: 104-81-4
EINECS: 203-240-6
UN: 3417
ADR: 6.1,II
Merck Index: 12,10223

Order Now

Specification Test Results

Appearance White to light yellow Crystalline mass
Infrared spectrum Conforms
Melting point 33 to 37 °C
GC ≥97.5 %

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR