tert-Butylamine 99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: 2-Aminoisobutane

107820010 107820050 107820025 107820100
ACRO107820010EA 43 EUR
ACRO107820010 ACRO107820050 ACRO107820025 ACRO107820100
tert-Butylamine 99%
tert-Butylamine
Formule: C₄H₁₁N
MW: 73.14 g/mol
Kookpunt: 46 °C (760 mmHg)
Smeltpunt: –67 °C
Dichtheid: 0.69 g/cm³
Vlampunt: –38 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008050
CAS nummer: 75-64-9
EINECS: 200-888-1
UN: 2734
ADR: 8 I
Merck Index: 13,01544

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.3770 to 1.3790 (20°C, 589 nm)
Water ≤0.3 %
GC ≥98.5 %
Color scale ≤10 APHA
SO4 and SO3 (as SO4) ≤0.005 %
Iron (Fe) ≤5 ppm
Heavy metals (as Pb) ≤5 ppm
Titrable acid ≤0.002 meq/g
Insoluble matter ≤0.005 %
Starch Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR