Butylbenzene 99+%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: n-Butylbenzene

107850500 107852500 107850010
ACRO107850500EA 52 EUR
ACRO107850500 ACRO107852500 ACRO107850010
Butylbenzene 99+%
Butylbenzene
Formule: C₁₀H₁₄
Kookpunt: 183 °C (760 mmHg)
Smeltpunt: –88 °C
Dichtheid: 0,86 g/cm³
Vlampunt: 59 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00009463
CAS nummer: 104-51-8
EINECS: 203-209-7
UN: 2709
ADR: 3,III
Merck Index: 13,01548

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4880 to 1.4900 (20°C, 589 nm)
GC ≥99.0 %
Methanol ≤0.5 % (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR