Ethyl-4-oxo-3-piperidinecarboxylate hydrochloride 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

108330050 108330250 108331000
ACRO108330050EA 47.3 EUR
ACRO108330050 ACRO108330250 ACRO108331000
Ethyl-4-oxo-3-piperidinecarboxylate hydrochloride 98%
Ethyl-4-oxo-3-piperidinecarboxylate hydrochloride

CAS No.: 4644-61-5

Formule: C₈H₁₃NO₃
Smeltpunt: 169…175 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00043267
CAS nummer: 4644-61-5

Order Now

Specification Test Results

Appearance White to yellow Crystalline powder
Infrared spectrum Conforms
Melting point 169 to 175 °C
Titration with HClO4 ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR