DL-Carnitine hydrochloride 99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

108450250 108451000
ACRO108450250EA 33.9 EUR
ACRO108450250 ACRO108451000
DL-Carnitine hydrochloride 99%
DL-Carnitine hydrochloride
Formule: C₇H₁₆ClNO₃
Smeltpunt: 190…205 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011904
CAS nummer: 461-05-2
EINECS: 207-309-1

Order Now

Specification Test Results

Appearance White to off-white Powder or crystals
Infrared spectrum Conforms
Melting point 194 to 200 °C
Specific optical rotation -1° to +1° (20 °C, 589 nm) (c=1, H2O)
Titration with HClO4 ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR