4-Chloor-o-toluidine 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: 2-Amino-5-chlorotoluene, 4-Chloro-2-methylaniline, 4-Chloro-o-toluidine, Fast Red TR Base

109370050 109371000
ACRO109370050EA 35 EUR
ACRO109370050 ACRO109371000
4-Chloor-o-toluidine 98%
4-Chloor-o-toluidine
Formule: C₇H₈ClN
MW: 141.6 g/mol
Kookpunt: 241
Smeltpunt: 29…30 °C
Dichtheid: 1.19 g/cm³
Vlampunt: 124 °C
MDL Nummer: MFCD00007842
CAS nummer: 95-69-2
EINECS: 202-441-6
UN: 2239
ADR: 6.1 III

Order Now

Specification Test Results

Appearance Clear brown Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.5845 to 1.5865 (20°C, 589 nm)
GC ≥97.5 %
Insoluble matter ≤0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR