2-Chloro-2-methylpropane 99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: tert-Butyl chloride

109500025 109500250 109505000
ACRO109500025EA 140 EUR
ACRO109500025 ACRO109500250 ACRO109505000
2-Chloro-2-methylpropane 99%
2-Chloro-2-methylpropane
Formule: C₄H₉Cl
Kookpunt: 51 °C (1013 hPa)
Smeltpunt: –25 °C
Dichtheid: 0,85 g/cm³
Vlampunt: –27 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00000816
CAS nummer: 507-20-0
EINECS: 208-066-4
UN: 1127
ADR: 3,II
Merck Index: 13,01561

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.3838 to 1.3858 (20°C, 589 nm)
GC ≥98.5 %
Color scale ≤15 APHA


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR