(-)-Cinchonidine 98.5-101% (dry basis)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

110330250 110331000
ACRO110330250EA 69.2 EUR
ACRO110330250 ACRO110331000
(-)-Cinchonidine 98.5-101% (dry basis)
(-)-Cinchonidine
Formule: C₁₉H₂₂N₂O
Kookpunt: 465 °C (1013 hPa)
Smeltpunt: 200…203 °C
Dichtheid: 1,22 g/cm³ (20 °C)
MDL Nummer: MFCD00006783
CAS nummer: 485-71-2
EINECS: 207-622-3
Merck Index: 13,02307

Order Now

Specification Test Results

Appearance White to cream Crystalline powder
Infrared spectrum Conforms
Melting point ≥200 °C
Loss on drying ≤1 % (105 °C)
Specific optical rotation -105° to -115° (20 °C, 589 nm) (c=1, EtOH) (on dried substance)
Titration with HClO4 98.5 to 101.0 % (On dried substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR