Nicotinonitrile 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

110860050 110861000 110865000 110860025
ACRO110860050EA 23.2 EUR
ACRO110860050 ACRO110861000 ACRO110865000 ACRO110860025
Nicotinonitrile 98%
Nicotinonitrile
Formule: C₆H₄N₂
Kookpunt: 206,2 °C (1013 hPa)
Smeltpunt: 47…51 °C
Dichtheid: 1,159 g/cm³
Vlampunt: 84 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00087130
CAS nummer: 100-54-9
EINECS: 202-863-0

Order Now

Specification Test Results

Appearance Colorless to white to yellow Crystals or crystalline mass and/or chunks
Infrared spectrum Conforms
Melting point 47 to 51 °C
Water ≤0.2 %
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR