1,4-Cyclohexanediol cis- and trans-mixture 99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

111150250 111151000
ACRO111150250EA 28.9 EUR
ACRO111150250 ACRO111151000
1,4-Cyclohexanediol cis- and trans-mixture 99%
1,4-Cyclohexanediol
Formule: C₆H₁₂O₂
Kookpunt: 150 °C (20 mmHg)
Smeltpunt: 98…102 °C
Dichtheid: 1,17 g/cm³ (21.85 °C)
Vlampunt: 65 °C
MDL Nummer: MFCD00001448
CAS nummer: 556-48-9
EINECS: 209-126-2

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 97 to 102 °C
GC ≥98.5 %
Iron (Fe) ≤20 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR