Cyclopentanon 99+%, pure

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

111530010 111532500 111530025 111530100
ACRO111530010EA 85.6 EUR
ACRO111530010 ACRO111532500 ACRO111530025 ACRO111530100
Cyclopentanon 99+%, pure
Cyclopentanon
Formule: C₅H₈O
MW: 84.12 g/mol
Kookpunt: 130…131 °C (760 mmHg)
Smeltpunt: –51 °C
Dichtheid: 0.95 g/cm³
Vlampunt: 26 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00001409
CAS nummer: 120-92-3
EINECS: 204-435-9
UN: 2245
ADR: 3 III
Merck Index: 13,02771

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4360 to 1.4380 (20°C, 589 nm)
GC ≥88 %
Stabilizer 0.01 % alpha-Tocopherol


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR