2,6-Dichloorfenol 99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

113590010 113590250 113591000
ACRO113590010EA 361 EUR
ACRO113590010 ACRO113590250 ACRO113591000
2,6-Dichloorfenol 99%
2,6-Dichloorfenol
Formule: C₆H₄Cl₂O
MW: 163 g/mol
Kookpunt: ∼219 °C (1013 hPa)
Smeltpunt: 65…68 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002176
CAS nummer: 87-65-0
EINECS: 201-761-3
UN: 2020
ADR: 6.1 III
Merck Index: 13,03099

Order Now

Specification Test Results

Appearance White to yellow Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 65 to 68 °C
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR