Dichloro(phenyl)phosphine 97%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

113661000 113665000
ACRO113661000EA 63.8 EUR
ACRO113661000 ACRO113665000
Dichloro(phenyl)phosphine 97%
Dichloro(phenyl)phosphine
Formule: C₆H₅Cl₂P
Kookpunt: 222 °C (1013 hPa)
Smeltpunt: –51 °C
Dichtheid: 1,319 g/cm³
Vlampunt: 102 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00000528
CAS nummer: 644-97-3
EINECS: 211-425-8
UN: 2798
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5970 to 1.5990 (20°C, 589 nm)
GC ≥96.0 %
Phosphate (PO4) ≤5 ppm
Calcium (Ca) ≤300 ppm
Iron (Fe) ≤5 ppm
Magnesium (Mg) ≤50 ppm
Potassium (K) ≤50 ppm
Heavy metals (as Pb) ≤5 ppm
Silicon dioxide (SiO2) ≤50 ppm
Sulfur compounds (as SO4) ≤0.004 %
Insoluble matter ≤0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR