4-(Dimethylamino)cinnamaldehyde 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

115790050 115790250
ACRO115790050EA 92.7 EUR
ACRO115790050 ACRO115790250
4-(Dimethylamino)cinnamaldehyde 98%
4-(Dimethylamino)cinnamaldehyde
Formule: C₁₁H₁₃NO
Smeltpunt: 137…142 °C
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD00007002
CAS nummer: 6203-18-5
EINECS: 228-267-0

Order Now

Specification Test Results

Appearance Yellow to orange Powder
Infrared spectrum Conforms
Melting point 137 to 142 °C
Titration with HClO4 ≥97.5 %
Stabilizer 0.1 % Hydroquinone


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR