Dimethylcarbonaat 99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

116120025 116125000 116120010 116120100
ACRO116120025EA 176 EUR
ACRO116120025 ACRO116125000 ACRO116120010 ACRO116120100
Dimethylcarbonaat 99%
Dimethylcarbonaat
Formule: C₃H₆O₃
MW: 90.08 g/mol
Kookpunt: 90 °C (760 mmHg)
Smeltpunt: 2…4 °C
Dichtheid: 1.069 g/cm³
Vlampunt: 18 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008420
CAS nummer: 616-38-6
EINECS: 210-478-4
UN: 1161
ADR: 3 II
Merck Index: 12,06117

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.3672 to 1.3692 (20°C, 589 nm)
Water ≤0.1 % (K.F.)
GC ≥98.5 %
Color scale ≤30 APHA
Acidity (HCl) ≤0.1 %
Methanol ≤0.2 % (GC)
Iron (Fe) ≤0.2 ppm
Heavy metals (as Pb) ≤0.5 ppm
Titrable base ≤0.0004 meq/g
KMnO4-reducing substance Passes test
K2Cr2O7-reducing substance Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR