Diphenylacetylene 99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

117180050 117180250 117181000
ACRO117180050EA 32.5 EUR
ACRO117180050 ACRO117180250 ACRO117181000
Diphenylacetylene 99%
Diphenylacetylene
Formule: C₁₄H₁₀
Kookpunt: 170 °C (19 mmHg)
Smeltpunt: 58…61 °C
Dichtheid: 0,99 g/cm³
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004786
CAS nummer: 501-65-5
EINECS: 207-926-6
Merck Index: 12,09643

Order Now

Specification Test Results

Appearance Light yellow to brown Crystalline powder and/or chunks
Infrared spectrum Conforms
Melting point 58 to 61 °C
GC ≥98.5 %
Calcium (Ca) ≤100 ppm
Magnesium (Mg) ≤50 ppm
Potassium (K) ≤50 ppm
Sodium (Na) ≤500 ppm
Insoluble matter ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR