Ferrocene 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

119141000 119145000 119140025
ACRO119141000EA 63 EUR
ACRO119141000 ACRO119145000 ACRO119140025
Ferrocene 98%
Ferrocene
Formule: C₁₀H₁₀Fe
Kookpunt: 249 °C (760 mmHg)
Smeltpunt: 173…176 °C
Dichtheid: 1,49 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00001427
CAS nummer: 102-54-5
EINECS: 203-039-3
UN: 1325
ADR: 4.1,II
Merck Index: 13,04070

Order Now

Specification Test Results

Appearance Orange-brown Crystalline powder or crystals (May become lumpy on storage)
Infrared spectrum Conforms
Melting point 173 to 176 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR