3-Fluorobenzaldehyde 98+%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

119330100 119330500 119332500
ACRO119330100EA 34.5 EUR
ACRO119330100 ACRO119330500 ACRO119332500
3-Fluorobenzaldehyde 98+%
3-Fluorobenzaldehyde
Formule: C₇H₅FO
Kookpunt: 66…68 °C (20 mmHg)
Dichtheid: 1,17 g/cm³
Vlampunt: 56 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003348
CAS nummer: 456-48-4
EINECS: 207-266-9
UN: 1989
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5170 to 1.5190 (20°C, 589 nm)
GC ≥98.0 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR