1-Fluoro-2-nitrobenzene 99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

119480100 119480500 119482500
ACRO119480100EA 20.9 EUR
ACRO119480100 ACRO119480500 ACRO119482500
1-Fluoro-2-nitrobenzene 99%
1-Fluoro-2-nitrobenzene
Formule: C₆H₄FNO₂
Kookpunt: 215 °C (760 mmHg)
Smeltpunt: 9…6 °C
Dichtheid: 1,335 g/cm³
Vlampunt: 94 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00007048
CAS nummer: 1493-27-2
EINECS: 216-088-0
UN: 2810
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Clear yellow to brown Liquid
Infrared spectrum Conforms
Refractive index 1.5307 to 1.5327 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR