2-Fluoropyridine 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

119630500 119630050
ACRO119630500EA 148 EUR
ACRO119630500 ACRO119630050
2-Fluoropyridine 98%
2-Fluoropyridine
Formule: C₅H₄FN
Kookpunt: 126 °C (753 mmHg)
Dichtheid: 1,12 g/cm³
Vlampunt: 24 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00006224
CAS nummer: 372-48-5
EINECS: 206-757-5
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4670 to 1.4690 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR