Fumaarzuur 99+%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

119750010 119750025 119751000 119750100
ACRO119750010EA 38.8 EUR
ACRO119750010 ACRO119750025 ACRO119751000 ACRO119750100
Fumaarzuur 99+%
Fumaarzuur
Formule: C₄H₄O₄
Kookpunt: 356 °C (1013 hPa)
Dichtheid: 1,62 g/cm³
Vlampunt: 230 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002700
CAS nummer: 110-17-8
EINECS: 203-743-0
Merck Index: 13,04308

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 295 to 300 °C
Water ≤0.25 % (K.F.)
HPLC ≥99.0 %
Titration with NaOH 99.0 to 101.0 % (On anhydrous substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR