Sodium-L(+)-glutamate monohydrate 99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: (S)-(+)-2-Aminopentanedioic acid sodium salt monohydrate, Sodium L-Glutamate Monohydrate, L(+)-Glutamic acid sodium salt monohydrate, L(+)-2-Aminopentanedioic acid sodium salt monohydrate, (S)-(+)-Glutamic acid sodium salt monohydrate, L(+)-Glutamic acid monosodium salt monohydrate, (S)-(+)-1-Aminopropane-1,3-dicarboxylic acid sodium salt monohydrate, (S)-(+)-Sodium glutamate monohydrate, L(+)-α-Glutamic acid sodium salt monohydrate, L(+)-1-Aminopropane-1,3-dicarboxylic acid sodium salt monohydrate, (S)-(+)-α-Glutamic acid sodium salt monohydrate

119940010 119940025 119940100
ACRO119940010EA 66.1 EUR
ACRO119940010 ACRO119940025 ACRO119940100
Sodium-L(+)-glutamate monohydrate 99%
Sodium-L(+)-glutamate monohydrate
Formule: C₅H₈NNaO₄·H₂O
Vlampunt: ≥ 225 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00150138
CAS nummer: 6106-04-3
EINECS: 240-313-1
Merck Index: 13,06278

Order Now

Specification Test Results

Appearance White to almost white Crystalline powder or crystals
Infrared spectrum Conforms
Loss on drying ≤0.5 % (100°C, 5 hrs)
Specific optical rotation +24.5° to +25.5° (20°C, 589 nm) (c=10, 2 N HCl)
Titration with HClO4 ≥98.5 %
Chloride (Cl) ≤2000 ppm
Sulfate (SO4) ≤500 ppm
Arsenic (As) ≤3 ppm
Heavy metals (as Pb) ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR