Glycinamide hydrochloride (H-Gly-NH2.HCl) 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

120060250 120061000
ACRO120060250EA 77.8 EUR
ACRO120060250 ACRO120061000
Glycinamide hydrochloride (H-Gly-NH2.HCl) 98%
Glycinamide hydrochloride (H-Gly-NH2.HCl)
Formule: C₂H₇ClN₂O
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00013008
CAS nummer: 1668-10-6
EINECS: 216-789-1

Order Now

Specification Test Results

Appearance White to beige Crystalline powder
Infrared spectrum Conforms
Titration Argentometric ≥97.5 %
Loss on drying ≤1 % (105°C, 4 hrs)
TLC ≤1 % Glycine


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR