4-Iodophenol 99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

122390100 122390250 122391000
ACRO122390100EA 53.7 EUR
ACRO122390100 ACRO122390250 ACRO122391000
4-Iodophenol 99%
4-Iodophenol
Formule: C₆H₅IO
Kookpunt: 287 °C (760 mmHg)
Smeltpunt: 89…94 °C
Dichtheid: 2,13 g/cm³ (25 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002327
CAS nummer: 540-38-5
EINECS: 208-745-5
UN: 3261
ADR: 8,III
Merck Index: 13,05058

Order Now

Specification Test Results

Appearance Beige to pink to brown Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 89 to 94 °C
GC ≥98.5 %

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR