3-Iodo-L-tyrosine 97%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

122440010 122440050 122440250
ACRO122440010EA 43 EUR
ACRO122440010 ACRO122440050 ACRO122440250
3-Iodo-L-tyrosine 97%
3-Iodo-L-tyrosine
Formule: C₉H₁₀INO₃
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00002608
CAS nummer: 70-78-0
EINECS: 200-744-8

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Water ≤4 % (K.F.)
Solubility (5 % in 1 N HCl) Clear colorless to light yellow
HPLC ≥96.0 %
Specific optical rotation -3° to -5° (20°C, 589 nm) (c=5 in 1 N HCl)

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR