DL-Malic acid 99+%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

125250010 125250050 125252500 125255000
ACRO125250010EA 65.4 EUR
ACRO125250010 ACRO125250050 ACRO125252500 ACRO125255000
DL-Malic acid 99+%
DL-Malic acid
Formule: HOOCCH(OH)CH₂COOH
Kookpunt: 306 °C (1013 hPa)
Smeltpunt: 128…132 °C
Dichtheid: 1,601 g/cm³ (20 °C)
Vlampunt: 203 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00064212
CAS nummer: 6915-15-7
EINECS: 210-514-9
Merck Index: 13,05730
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=920b54632a869bc5df22624fb9fecd4d11cae65e&vid=278193678126274041defdc0bd71c7c40efb3085&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 128 to 132 °C
Sulfated ash ≤0.1 %
Specific optical rotation -0.1° to +0.1° (25 °C, 589 nm) (C=8.5%,H2O)
Titration with NaOH ≥99.0 %
Lead (Pb) ≤2 ppm
Impurity ≤0.05 % Maleic acid
Insoluble matter ≤0.1 % (in water)

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR