Malonic acid 99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: 1,3-Propanedioic acid

125260010 125262500 125260051
ACRO125260010EA 262 EUR
ACRO125260010 ACRO125262500 ACRO125260051
Malonic acid 99%
Malonic acid
Formule: C₃H₄O₄
MW: 104.06 g/mol
Kookpunt: 387
Smeltpunt: 130…135 °C (ontbinding)
Vlampunt: 172 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002707
CAS nummer: 141-82-2
EINECS: 205-503-0
Merck Index: 13,05732

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 133 to 136 °C
Titration with NaOH ≥98.5 %
Chloride (Cl) ≤500 ppm
Sulfate (SO4) ≤500 ppm

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR