Methylchloorformiaat 99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

126530050 126535000 126531000 126530025
ACRO126530050EA 38.4 EUR
ACRO126530050 ACRO126535000 ACRO126531000 ACRO126530025
Methylchloorformiaat 99%
Methylchloorformiaat
Formule: C₂H₃ClO₂
Kookpunt: 70…72 °C (760 mmHg)
Smeltpunt: –61 °C
Dichtheid: 1,223 g/cm³
Vlampunt: 5 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00000639
CAS nummer: 79-22-1
EINECS: 201-187-3
UN: 1238
ADR: 6.1,
Merck Index: 12,06123

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.3860 to 1.3880 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR