Methyl-4-hydroxybenzoate 99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Methylparaben, 4-Hydroxybenzoic acid methyl ester, Methyl Parahydroxybenzoate, Methyl 4-hydroxybenzoate

126961000 126965000 126960025 126960100
ACRO126961000EA 33.7 EUR
ACRO126961000 ACRO126965000 ACRO126960025 ACRO126960100
Methyl-4-hydroxybenzoate 99%
Methyl-4-hydroxybenzoate
Formule: HOC₆H₄CO₂CH₃
MW: 152.15 g/mol
Kookpunt: 298.6 °C (760 mmHg)
Smeltpunt: 125…128 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002352
CAS nummer: 99-76-3
EINECS: 202-785-7
Merck Index: 13,06129
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=84321052052f4be2a53d8d7ebac7fabe6fc5a435&vid=ede765fad275b2515a8c57ba4499bff313a18983&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to almost white Crystalline powder
Infrared spectrum Conforms
Melting point 125 to 128 °C
HPLC ≥98.5 %
Residue after ignition ≤0.1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR