3-Nitro-p-toluidine 97%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: 3-Nitro-4-methylaniline, 4-Methyl-3-nitroaniline, 4-Amino-2-nitrotoluene

127250250 127250050 127251000
ACRO127250250EA 47.9 EUR
ACRO127250250 ACRO127250050 ACRO127251000
3-Nitro-p-toluidine 97%
3-Nitro-p-toluidine
Formule: C₇H₈N₂O₂
MW: 152.15 g/mol
Kookpunt: 306 °C (1013 hPa)
Smeltpunt: 74…78 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00007910
CAS nummer: 119-32-4
EINECS: 204-314-0
UN: 2660
ADR: 6.1 III

Order Now

Specification Test Results

Appearance Ochre to orange to brown Powder
Infrared spectrum Conforms
Melting point 74 to 78 °C
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR