2-Naphthylacetic acid 99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

128220050 128220250 128221000
ACRO128220050EA 42.2 EUR
ACRO128220050 ACRO128220250 ACRO128221000
2-Naphthylacetic acid 99%
2-Naphthylacetic acid
Formule: C₁₂H₁₀O₂
Kookpunt: 352 °C (1013 hPa)
Smeltpunt: 141…143 °C
Dichtheid: 1,29 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004126
CAS nummer: 581-96-4
EINECS: 209-475-0

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder or flakes
Infrared spectrum Conforms
Melting point 141 to 143 °C
GC ≥98.5 %
Titration with NaOH ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR