Nicotinamide 99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

128271000 128275000 128270025
ACRO128271000EA 36.8 EUR
ACRO128271000 ACRO128275000 ACRO128270025
Nicotinamide 99%
Nicotinamide
Formule: C₆H₆N₂O
Kookpunt: 150…160 °C (760 mmHg)
Smeltpunt: 128…131 °C
Dichtheid: 1,4463 g/cm³ (20 °C)
Vlampunt: >150 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00006395
CAS nummer: 98-92-0
EINECS: 202-713-4
Merck Index: 12,06574

Order Now

Specification Test Results

Appearance White to almost white Crystalline powder
Infrared spectrum Conforms
Melting point 128 to 131 °C
Loss on drying ≤0.5 % (4 hrs) (silica gel)
Sulfated ash ≤0.1 %
Readily carbonizable substance Passes test
pH 6.0 to 7.5 (5 % at 20 °C)
Titration with HClO4 ≥98.5 % (on dried substance)
Heavy metals (as Pb) ≤30 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR