Phenylphosphonic acid 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

130791000 130795000 130790025
ACRO130791000EA 49.9 EUR
ACRO130791000 ACRO130795000 ACRO130790025
Phenylphosphonic acid 98%
Phenylphosphonic acid
Formule: C₆H₇O₃P
Smeltpunt: 158…165 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002136
CAS nummer: 1571-33-1
EINECS: 216-388-1
UN: 3261
ADR: 8,III

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 158 to 165 °C
Titration with NaOH ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR