4-Picolyl chloride hydrochloride 97%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

131200100 131201000
ACRO131200100EA 44.4 EUR
ACRO131200100 ACRO131201000
4-Picolyl chloride hydrochloride 97%
4-Picolyl chloride hydrochloride
Formule: C₆H₇Cl₂N
Smeltpunt: 160…163 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00012826
CAS nummer: 1822-51-1
EINECS: 217-350-7
UN: 3261
ADR: 8,III

Order Now

Specification Test Results

Appearance Yellow to orange Crystalline powder
Infrared spectrum Conforms
Titration with HClO4 ≥96.0 %
GC ≥99.4 %
Color scale ≤20 APHA


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR