(1S)-(-)-α-Pinene 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

131270050 131272500 131275000 131270025
ACRO131270050EA 19.3 EUR
ACRO131270050 ACRO131272500 ACRO131275000 ACRO131270025
(1S)-(-)-α-Pinene 98%
(1S)-(-)-α-Pinene
Formule: C₁₀H₁₆
Kookpunt: 155…156 °C (760 mmHg)
Smeltpunt: –64 °C
Dichtheid: 0,8595 g/cm³
Vlampunt: 33 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00064145
CAS nummer: 7785-26-4
EINECS: 232-077-3
UN: 2368
ADR: 3,III
Merck Index: 12,07599

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4640 to 1.4680 (20°C, 589 nm)
GC ≥97.5 %
Color scale ≤30 APHA
Specific gravity (20°C) 0.858 to 0.861
Specific optical rotation -35° to -45° (20°C, 589 nm) (Neat)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR