Piperazine hexahydrate 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

131300010 131301000 131305000 131300051
ACRO131300010EA 120 EUR
ACRO131300010 ACRO131301000 ACRO131305000 ACRO131300051
Piperazine hexahydrate 98%
Piperazine hexahydrate
Formule: C₄H₁₀N₂·6H₂O
Kookpunt: 145…156 °C (760 mmHg)
Smeltpunt: 41…45 °C
Vlampunt: 87 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149389
CAS nummer: 142-63-2
EINECS: 203-808-3
UN: 2579
ADR: 8,III
Merck Index: 12,07617

Order Now

Specification Test Results

Appearance Transparent to white Adhering crystals
Infrared spectrum Conforms
Titration with HCl ≥97.5 %
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR