Sodium pyruvate 99+%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: 2-Ketopropionic acid sodium salt, 2-Oxopropionic acid sodium salt, Sodium 2-oxopropanoate, Pyruvic acid sodium salt, α-Ketopropionic acid sodium salt

132150250 132151000 132155000
ACRO132150250EA 39.4 EUR
ACRO132150250 ACRO132151000 ACRO132155000
Sodium pyruvate 99+%
Sodium pyruvate
Formule: CH₃COCOONa
Smeltpunt: >300 °C
Dichtheid: 1,718 g/cm³ (20 °C)
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00002586
CAS nummer: 113-24-6
EINECS: 204-024-4

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤0.5 %
Heavy metals ≤10 ppm
Solubility (10 % in water) Clear colorless to light yellow
Titration with HClO4 ≥99.0 %
Chloride (Cl) ≤20 ppm
Sulfate (SO4) ≤20 ppm
Arsenic (As) ≤1 ppm

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR