Quinaldine 95+%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: 2-Methylquinoline

132172500 132170010
ACRO132172500EA 77.8 EUR
ACRO132172500 ACRO132170010
Quinaldine 95+%
Quinaldine
Formule: C₁₀H₉N
Kookpunt: 248 °C (760 mmHg)
Smeltpunt: –2 °C
Dichtheid: 1,058 g/cm³
Vlampunt: 79 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00006756
CAS nummer: 91-63-4
EINECS: 202-085-1
Merck Index: 12,08228

Order Now

Specification Test Results

Appearance Clear light yellow to brown-yellow Liquid
Infrared spectrum Conforms
Refractive index 1.6080 to 1.6130 (20°C, 589 nm)
GC ≥95.0 %
Hydroxyl value 25 to 32 mg KOH/g

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR