β-Sitosterol with ca. 10% campesterol, ca. 75% beta-sitosterol

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

132720050 132721000 132725000
ACRO132720050EA 33.6 EUR
ACRO132720050 ACRO132721000 ACRO132725000
β-Sitosterol with ca. 10% campesterol, ca. 75% beta-sitosterol
beta-Sitosterol
Formule: C₂₉H₅₀O
Smeltpunt: 139…142 °C
Vlampunt: >100 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00003631
CAS nummer: 83-46-5

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Loss on drying ≤3.5 %
GC ≤3.0 % (Others) Typical
Specific optical rotation -24° to -32° (20°C, 589 nm) (c=2, CHCl3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR