4-Iodotoluene 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

133940100 133940500 133941000
ACRO133940100EA 37.4 EUR
ACRO133940100 ACRO133940500 ACRO133941000
4-Iodotoluene 98%
4-Iodotoluene
Formule: C₇H₇I
Kookpunt: 211,5 °C (1013 hPa)
Smeltpunt: 33…36 °C
Dichtheid: 1,68 g/cm³ (20 °C)
Vlampunt: 90 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00001059
CAS nummer: 624-31-7
EINECS: 210-841-7

Order Now

Specification Test Results

Appearance White to light yellow Low melting crystalline mass
Infrared spectrum Conforms
Melting point 33 to 36 °C
GC ≥97.5 %
Elemental Analysis 7.55 to 8.35 % N


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR