5-Aminosalicylic acid 99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

134330050 134330250 134331000
ACRO134330050EA 24.5 EUR
ACRO134330050 ACRO134330250 ACRO134331000
5-Aminosalicylic acid 99%
5-Aminosalicylic acid
Formule: C₇H₇NO₃
Kookpunt: 347 °C (1013 hPa)
Dichtheid: 1,568 g/cm³ (20 °C)
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00007877
CAS nummer: 89-57-6
EINECS: 201-919-1
Merck Index: 12,05964

Order Now

Specification Test Results

Appearance Beige to grey to light brown Powder
Infrared spectrum Conforms
Loss on drying ≤0.5 % (1 g, 105°C, 3 hrs)
HPLC ≥98.5 %
Titration with HClO4 ≥98.5 % (On dry substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR