D-(-)-Tartaric acid 99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

137860250 137861000 137862500 137860010
ACRO137860250EA 55 EUR
ACRO137860250 ACRO137861000 ACRO137862500 ACRO137860010
D-(-)-Tartaric acid 99%
D-(-)-Tartaric acid
Formule: C₄H₆O₆
Kookpunt: 356 °C (1013 hPa)
Smeltpunt: 166…170 °C
Dichtheid: 1,76 g/cm³ (20 °C)
Vlampunt: 210 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004238
CAS nummer: 147-71-7
EINECS: 205-695-6
Merck Index: 12,09235

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 167 to 171 °C
Loss on drying ≤0.5 % (1 g, 105 °C, 3 hrs)
Sulfated ash ≤0.1 %
Specific optical rotation -11.0° to -13.0° (20 °C, 589 nm) (c=20, water)
Titration with NaOH ≥98.5 %
Heavy metals (as Pb) ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR