Triacetin (glycerol triacetate) 99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Glyceryl triacetate, Propane-1,2,3-triyl triacetate, Glycerol triacetate

139220010 139220025 139225000 139220100 139220000
ACRO139220010EA 62.2 EUR
ACRO139220010 ACRO139220025 ACRO139225000 ACRO139220100 ACRO139220000
Triacetin (glycerol triacetate) 99%
Triacetin (glycerol triacetate)
Formule: C₉H₁₄O₆
MW: 218.21 g/mol
Kookpunt: 258 °C (760 mmHg)
Smeltpunt: 3 °C
Dichtheid: 1.155 g/cm³
Vlampunt: 138 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008716
CAS nummer: 102-76-1
EINECS: 203-051-9
Merck Index: 13,09664

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4290 to 1.4330 (20°C, 589 nm)
Density (25°C) 1.154 to 1.158 g/ml
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR