Triethylboraat 97%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

139582500 139580010
ACRO139582500EA 84 EUR
ACRO139582500 ACRO139580010
Triethylboraat 97%
Triethylboraat
Formule: C₆H₁₅BO₃
MW: 145.99 g/mol
Kookpunt: 117…118 °C (760 mmHg)
Smeltpunt: –84.5 °C
Dichtheid: 0.858 g/cm³
Vlampunt: 11 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00012846
CAS nummer: 150-46-9
EINECS: 205-760-9
UN: 1176
ADR: 3 II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3705 to 1.3775 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR