Tris-(hydroxymethyl)-aminomethaan (TRIS) 99+% voor de biochemie

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

140500010 140500025 140505000 140500100
ACRO140500010EA 167 EUR
ACRO140500010 ACRO140500025 ACRO140505000 ACRO140500100
Tris-(hydroxymethyl)-aminomethaan (TRIS) 99+% voor de biochemie
Tris-(hydroxymethyl)-aminomethaan (TRIS)
Formule: NH₂C(CH₂OH)₃
Kookpunt: 219…220 °C (10 mmHg)
Dichtheid: 1,328 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004679
CAS nummer: 77-86-1
EINECS: 201-064-4
Merck Index: 13,09842
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=11a03d1ab6b2c11b9a2c83f0a73fc4264cec0110&vid=5ce890cdf298a59a7900a07fee1c857a5e5e61e0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 168 to 171 °C
Water ≤0.5 % (K.F.)
UV at 290 nm A: ≤0.2
Residue after ignition ≤0.1 %
pH 10.0 to 11.5 (5 % in water)
Titration with HClO4 ≥99.0 %
Color of solution ≤25 APHA (20 % in water)
Heavy metals (as Pb) ≤5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR