DL-m-Tyrosine 98.5%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

140630010 140630050
ACRO140630010EA 109 EUR
ACRO140630010 ACRO140630050
DL-m-Tyrosine 98.5%
DL-m-Tyrosine
Formule: C₉H₁₁NO₃
Kookpunt: 351 °C (1013 hPa)
Dichtheid: 1,456 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002597
CAS nummer: 775-06-4
EINECS: 212-270-9
Merck Index: 12,09971

Order Now

Specification Test Results

Appearance White to light beige Crystalline powder
Infrared spectrum Conforms
Water ≤0.5 % (K.F.)
Titration with HClO4 ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR