Vanillin 99%, pure

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: 4-Hydroxy-3-methoxybenzaldehyde

140820010 140820020 140821000 140822500 140820050
ACRO140820010EA 154 EUR
ACRO140820010 ACRO140820020 ACRO140821000 ACRO140822500 ACRO140820050
Vanillin 99%, pure
Vanillin
Formule: C₈H₈O₃
MW: 152.15 g/mol
Kookpunt: 170 °C (15 mmHg)
Smeltpunt: 81…84 °C
Vlampunt: 147 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00006942
CAS nummer: 121-33-5
EINECS: 204-465-2
Merck Index: 13,09998
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd6583c5aea50cd090e103650a05eab491b4d38e&vid=535f9a4c80d51d7343126ae5e79c6c83b3beefa7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 81 to 84 °C
Titration with NaOH ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR